Anti-FGA Goat Polyclonal Antibody
EVBIEB12376
- Antibody type:Primary
- Antigen name:Fibrinogen alpha chain
- Antigen symbol:FGA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Format:affinity purified
- Form:liquid
- Gene ID:2243
- Antigen synonyms:Fib2
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Sequence:C-STSYNRGDSTFES
- Storage temperature:Store at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing
- Concentration:100µg/200µl
- Shipping temperature:8 °C
- Purification:Affinity purified
- Size:100µg
- Pack type:plastic vial
- Pk:0,1 mg
Western Blot: Approx 75kDa band observed in Human Platelets lysates (calculated MW of 69.8kDa according to NP_068657.1). Recommended concentration: 0.01-0.03µg/ml.
Type: Primary
Antigen: FGA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human