Anti-ATG4A Goat Polyclonal Antibody
EVBIEB08867
- Antibody type:Primary
- Antigen name:autophagy related 4A, cysteine peptidase
- Antigen symbol:ATG4A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Format:affinity purified
- Form:liquid
- Gene ID:115201
- Antigen synonyms:APG4A|AUTL2
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Sequence:TEENGTVNDQTFHC
- Storage temperature:Store at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing
- Concentration:100µg/200µl
- Shipping temperature:8 °C
- Purification:Affinity purified
- Size:100µg
- Pack type:plastic vial
- Pk:0,1 mg
Type: Primary
Antigen: ATG4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human