Order Entry
Slovakia
ContactUsLinkComponent
Anti-CDK5 Mouse Monoclonal Antibody [clone: 1321CT281.130.129]
Anti-CDK5 Mouse Monoclonal Antibody [clone: 1321CT281.130.129]
Catalog # ABGEAM8417B
Supplier:  ABGENT
Anti-CDK5 Mouse Monoclonal Antibody [clone: 1321CT281.130.129]
Catalog # ABGEAM8417B
Supplier:  ABGENT

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    cyclin-dependent kinase 5
  • Antigen symbol:
    CDK5
  • Clonality:
    Monoclonal
  • Clone:
    1321CT281.130.129
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Gene ID:
    1020
  • Antigen synonyms:
    PSSALRE|LIS7
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    33.304 kDa
  • Sequence:
    AEVVTLWYRPPDVLFGAKLYSTSIDMWSAGCIFAELANAGRPLFPGNDVDDQLKRIFRLLGTPTEEQWPSMTKLPDYKPYPMYPATTSLVNVVPKLNATGRDLLQNLLKCNPVQRISAEEALQHPYF
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    1.450
  • Shipping temperature:
    4 °C
  • Immunogen:
    This antibody is generated from a mouse immunized with a recombinant protein from human.
  • Purification:
    Protein G Purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: CDK5
Clonality: Monoclonal
Clone: 1321CT281.130.129
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat