Order Entry
Slovakia
ContactUsLinkComponent
Anti-RAD23B Mouse Monoclonal Antibody [clone: 1228CT409.120.123.135]
Anti-RAD23B Mouse Monoclonal Antibody [clone: 1228CT409.120.123.135]
  ABGEAM2267B
 :  ABGENT
undefined
Anti-RAD23B Mouse Monoclonal Antibody [clone: 1228CT409.120.123.135]
  ABGEAM2267B
 :  ABGENT

 

  • Antibody type:
    Primary
  • Antigen name:
    RAD23 homolog B
  • Antigen symbol:
    RAD23B
  • Clonality:
    Monoclonal
  • Clone:
    1228CT409.120.123.135
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Gene ID:
    5887
  • Antigen synonyms:
    HR23B|HHR23B|P58
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    43.171 kDa
  • Sequence:
    MQVTLKTLQQQTFKIDIDPEETVKALKEKIESEKGKDAFPVAGQKLIYAGKILNDDTALKEYKIDEKNFVVVMVTKPKAVSTPAPATTQQSAPASTTAVTSSTTTTVAQAPTPVPALAPTSTPASITPASATASSEPAPASAAKQEKPAEKPAETPVATSPTATDSTSGDSSRSNLFEDATSALVTGQSYENMVTEIMSMGYEREQVIAALRASFNNPDRAVEYLLMGIPGDRESQAVVDPPQAASTGAPQSSAVAAAAATTTATTTTTSSGGHPLEFLRNQPQFQQMRQIIQQNPSLLPALLQQIGRENPQLLQQISQHQEHFIQMLNEPVQEAGGQGGGGGGGSGGIAEAGSGHMNYIQVTPQEKEAIERLKALGFPEGLVIQAYFACEKNENLAANFLLQQNFDED
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.5
  • Shipping temperature:
    4 °C
  • Immunogen:
    This RAD23B antibody is generated from a mouse immunized with a KLH conjugated synthetic peptide between 1-409 amino acids from the N-terminal region of human RAD23B.
  • Purification:
    Protein G Purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: RAD23B
Clonality: Monoclonal
Clone: 1228CT409.120.123.135
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat