Order Entry
Slovakia
ContactUsLinkComponent
Anti-ING4 Rabbit Polyclonal Antibody
Anti-ING4 Rabbit Polyclonal Antibody
  ABGEAP20568A
 :  ABGENT
undefined
Anti-ING4 Rabbit Polyclonal Antibody
  ABGEAP20568A
 :  ABGENT

 

  • Antibody type:
    Primary
  • Antigen name:
    Inhibitor of Growth Family, Member 4
  • Antigen symbol:
    ING4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Gene ID:
    51147
  • Antigen synonyms:
    my036|p29ING4
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    28.53 kDa
  • Sequence:
    HPSDVLDMPVDPNE
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.62
  • Shipping temperature:
    4 °C
  • Immunogen:
    This ING4 antibody is generated from a rabbit immunized with a KLH conjugated synthetic peptide between 162-195 amino acids from the C-terminal region of human ING4.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: ING4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human