Distillation flasks, with side arm
Supplier: KAVALIER
Jump to:
About this item
Borosilicate glass 3.3, SIMAX®.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Flasks with side arm sealed at the bottom third part of neck. Uniform wall thickness distribution makes these flasks ideal for heating applications and distillations.
4 Options Available Below
Product Details & Documents
Borosilicate glass 3.3, SIMAX®.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Flasks with side arm sealed at the bottom third part of neck. Uniform wall thickness distribution makes these flasks ideal for heating applications and distillations.
Product Family Options
Product Information
- CapacityHeightExt.ØNeck I-ØAvailabilityPrice