Order Entry
Slovakia
ContactUsLinkComponent
Anti-ORM2 Rabbit Polyclonal Antibody
Anti-ORM2 Rabbit Polyclonal Antibody
Catalog # BOSSBS-7565R
Supplier:  BIOSS INC
Anti-ORM2 Rabbit Polyclonal Antibody
Catalog # BOSSBS-7565R
Supplier:  BIOSS INC

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Orosomucoid 2
  • Antigen symbol:
    ORM2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    5005
  • Antigen synonyms:
    AGP2|AGP-B'|AGP-B
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

This gene encodes a key acute phase plasma protein. Because of its increase due to acute inflammation, this protein is classified as an acute-phase reactant. The specific function of this protein has not yet been determined; however, it may be involved in aspects of immunosuppression. [provided by RefSeq, Jul 2008].

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ORM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human