Anti-STRO1 Mouse Monoclonal Antibody [clone: STRO-1]
ABNOMAB10247
: ABNOVA
- Antibody type:Primary
- Antigen symbol:STRO1
- Clonality:Monoclonal
- Clone:STRO-1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgM
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:STRO-1
- Storage buffer:TBS, pH 8.0 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:Human CD34 positive bone marrow cells.
- Purification:Precipitation methods and ion exchange chromatography
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against of STRO-1.
Type: Primary
Antigen: STRO1
Clonality: Monoclonal
Clone: STRO-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human