Anti-CEA Mouse Monoclonal Antibody [clone: II135F10 C4]
Catalog # ABNOMAB3675
Supplier: ABNOVA
Specifications
- Antibody type:Primary
- Antigen name:Carcinoembryonic Antigen
- Antigen symbol:CEA
- Clonality:Monoclonal
- Clone:II135F10 C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:CD66e|meconium antigen 100|Carcinoembryonic Antigen
- Storage buffer:BBS
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified human CEA.
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Mouse monoclonal antibody raised against native CEA.
Type: Primary
Antigen: CEA
Clonality: Monoclonal
Clone: II135F10 C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human