Order Entry
Slovakia
ContactUsLinkComponent
Rat IgG2a Isotype Control [clone: 2A3] (redFluor® 710)
Rat IgG2a Isotype Control [clone: 2A3] (redFluor® 710)
Catalog # 80-4321-U100
Supplier:  TONBO BIOSCIENCE
Rat IgG2a Isotype Control [clone: 2A3] (redFluor® 710)
Catalog # 80-4321-U100
Supplier:  TONBO BIOSCIENCE

Specifications

  • Antibody type:
    Control
  • Clone:
    2A3
  • Conjugation:
    redFluor® 710
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a
  • Neutralisation:
    Yes
  • Storage temperature:
    Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

The 2A3 immunoglobulin is useful as an isotype-matched control. The 2A3 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2a antibodies.

Type: Control
Antigen: 2A3
Clonality:
Clone: 2A3
Conjugation: redFluor™ 710
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: N/A