Order Entry
Slovakia
ContactUsLinkComponent
Anti-Mt1 Rabbit Polyclonal Antibody [clone: KH-1]
Anti-Mt1 Rabbit Polyclonal Antibody [clone: KH-1]
Supplier:  CAYMAN
Certificates
Anti-Mt1 Rabbit Polyclonal Antibody [clone: KH-1]
Supplier:  CAYMAN

Specifications

  • Catalog No:
  • CAYM172130-500
  • CAYM172130-100
  • Antigen symbol:
  • MT1
  • MT1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • KH-1
  • KH-1
  • Reactivity:
  • Rajiform sp., Yellow gurnard, Salmoniform, Cypriniform, Whiting, Atlantic cod, Flounder, Perciform, Siluriform
  • Rajiform sp., Yellow gurnard, Salmoniform, Cypriniform, Whiting, Atlantic cod, Flounder, Perciform, Siluriform
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • metallothionein from liver of Atlantic cod (Gadus morhua)
  • metallothionein from liver of Atlantic cod (Gadus morhua)
  • Purification:
  • purified
  • purified
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
  • 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
  • Storage temperature:
  • +4 °C
  • +4 °C
  • Shipping temperature:
  • 4 °C
  • 4 °C

Specifications

Frequently Bought Together