Anti-Bnc 1 Rabbit Polyclonal Antibody
Catalog # CAYM13502-1
Supplier: CAYMAN
Specifications
- Antibody type:Primary
- Antigen symbol:BNC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Chimpanzee,Human
- Western blot:Yes
- Cross adsorption:No
- Amino acid number:330 to 380
- Storage buffer:PBS containing 0,05 BSA and 0,05% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:synthetic peptide from human Bnc 1 within the region of amino acids 330-380
- Purification:purified by Protein G chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: Bnc 1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chimpanzee, Human, Monkey