Anti-NR2B subunit of NMDA Rabbit Polyclonal Antibody
CAYM10609-1
: CAYMAN
- Antibody type:Primary
- Antigen name:NR2B subunit of NMDA
- Antigen symbol:NR2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:fusion protein from the C-terminus of the NR2B subunit of rat NMDA receptor
- Purification:purified by affinity chromatography
- Size:1 Ea
- Pk:1
Type: Primary
Antigen: NR2B subunit of NMDA
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat