Order Entry
Slovakia
ContactUsLinkComponent
Anti-Syn1 Rabbit Polyclonal Antibody
Anti-Syn1 Rabbit Polyclonal Antibody
Catalog # CAYM10606-1
Supplier:  CAYMAN
Anti-Syn1 Rabbit Polyclonal Antibody
Catalog # CAYM10606-1
Supplier:  CAYMAN

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Synapsin 1
  • Antigen symbol:
    SYN1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    native protein from bovine brain
  • Purification:
    purified by affinity chromatography
  • Size:
    1 Ea
  • Pk:
    1

Specifications

About this item

Type: Primary
Antigen: Syn1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat