Anti-IgA Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # CAYM10196-500
Supplier: CAYMAN
Specifications
- Antibody type:Primary
- Antigen symbol:IgA
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Cross adsorption:No
- Storage buffer:PBS, pH 7,2, containing 50% glycerol, 0,1% BSA, and 0,02% sodium azide
- Storage temperature:+4 °C
- Shipping temperature:4 °C
- Immunogen:mouse IgA
- Purification:purified by Protein A chromatography
- Size:500 µl
- Pk:500 µl
Specifications
About this item
Type: Primary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse