Anti-NMDA Receptor NR2B Subunit Rabbit Polyclonal Antibody
Catalog # CAYM10009760-1
Supplier: CAYMAN
Specifications
- Antibody type:Primary
- Antigen name:NMDA Receptor NR2B subunit
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Modification:phospho-Tyr1336
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:phosphopeptide corresponding to amino acid residues surrounding phospho-Tyr1336 of the human NMDA Receptor NR2B Subunit
- Purification:purified by affinity chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: NMDA Receptor NR2B Subunit
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat