Anti-sRBP4 Rabbit Polyclonal Antibody
CAYM10007681-500
: CAYMAN
- Antibody type:Primary
- Antigen symbol:sRBP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Amino acid number:28 to 37
- Sequence:RVKENFDKAR
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human sRBP4 amino acids 28-37 (RVKENFDKAR)
- Purification:purified by affinity chromatography
- Size:500 µl
- Pk:500 µl
Type: Primary
Antigen: sRBP4
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human