Anti-ATGL Rabbit Polyclonal Antibody
CAYM10006409-1
: CAYMAN
- Antibody type:Primary
- Antigen name:Adipose Triglyceride Lipase
- Antigen symbol:ATGL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:lyophilized
- Amino acid number:382 to 400
- Storage buffer:TBS, pH 7,4
- Sequence:KRKLGRHLPSRLPEQVELR
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human adipose triglyceride lipase (ATGL) amino acids 382-400
- Purification:purified by affinity chromatography
- Size:1 Ea
- Pk:1
Type: Primary
Antigen: ATGL
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat