Order Entry
Slovakia
ContactUsLinkComponent
Anti-BIRC7 Mouse Monoclonal Antibody (Unconjugated) [clone: 88C570]
Anti-BIRC7 Mouse Monoclonal Antibody (Unconjugated) [clone: 88C570]
Catalog # ROCK200-301-H90
Supplier:  ROCKLAND
undefined
Anti-BIRC7 Mouse Monoclonal Antibody (Unconjugated) [clone: 88C570]
Catalog # ROCK200-301-H90
Supplier:  ROCKLAND

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    baculoviral IAP repeat containing 7
  • Antigen symbol:
    BIRC7
  • Clonality:
    Monoclonal
  • Clone:
    88C570
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    79444
  • Antigen synonyms:
    MLIAP|LIVIN|RNF50|ML-IAP|KIAP
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,05% (w/v) Sodium Azide
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Dry Ice
  • Immunogen:
    Livin Antibody was produced in mice prepared by repeated immunizations with recombinant human protein, Livin.
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Anti-Livin antibody is suitbale for use in IF/ICC and WB. Expect a band approximately 30kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Immunofluorescence: 2-5 µg/ml; Western Blot: 1-2 µg/ml

Type: Primary
Antigen: BIRC7
Clonality: Monoclonal
Clone: 88C570
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human