Anti-OSPA Rabbit Polyclonal Antibody
ROCK200-401-C13
: ROCKLAND
- Antibody type:Primary
- Antigen symbol:OSPA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:B. burgdorferi
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Gene ID:1194357
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:MBP-fusion Protein corresponding to Borrelia burgdorferi OspA Protein.
- Purification:Protein A purified
- Size:100 µg
- Pk:100 µG
This Protein-A purified antibody has been tested for use in Western blotting. Specific conditions for reactivity should be optimized by the user. Expect a band approximately 28.1 kDa in size corresponding to Borrelia burgdorferi OspA Protein by western blotting.
Recommended Dilutions: ELISA: >1:5000; Western Blot: 1:1000;
Type: Primary
Antigen: OSPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Borrelia burgdorferi