Order Entry
Slovakia
ContactUsLinkComponent
Anti-SLC1A6 Rabbit Polyclonal Antibody
Anti-SLC1A6 Rabbit Polyclonal Antibody
Catalog # ORIGTA322676
Dodávateľ:  ORIGENE TECHNOLOGIES
Anti-SLC1A6 Rabbit Polyclonal Antibody
Catalog # ORIGTA322676
Dodávateľ:  ORIGENE TECHNOLOGIES

Špecifikácie

  • Antibody type:
    Primary
  • Antigen symbol:
    Solute carrier family 1, member 6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Amino acid number:
    205 to 220
  • Storage buffer:
    PBS pH 7,3, 0,05% NaN3, 50% glycerol
  • Molecular weight:
    62 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Synthetic peptide corresponding to a region derived from 205-220 amino acids of Human Solute carrier family 1 member 6
  • Purification:
    purified by antigen affinity chromatography
  • Size:
    100 μl
  • Pk:
    100 µl

Špecifikácie

O tomto produkte

Recommended Dilutions: ELISA: 1:1000-5000; Western Blot: 1:200-1000

Type: Primary
Antigen: SLC1A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat