Anti-PDE1A Mouse Monoclonal Antibody [clone: 7D5]
Catalog # ORIGCF803629
Supplier: ORIGENE TECHNOLOGIES
Specifications
- Antibody type:Primary
- Antigen name:Phosphodiesterase 1A, calmodulin-dependent
- Antigen symbol:PDE1A
- Clonality:Monoclonal
- Clone:7D5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:259 to 545
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 259-545 of human PDE1A (NP_005010) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: PDE1A
Clonality: Monoclonal
Clone: 7D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human