Anti-DCN Rabbit Polyclonal Antibody
Catalog # ORIGTA318941
Supplier: ORIGENE TECHNOLOGIES
Specifications
- Antibody type:Primary
- Antigen symbol:DCN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Pig,Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:100 µg (0,5 mg/ml) affinity purified rabbit anti-Decorin polyclonal antibody in phosphate buffered saline (PBS), pH 7,2, containing 25% glycerol, 0,5% BSA, and 0,01% thimerosal.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Synthetic peptide surrounding amino acid 130 of human Decorin
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 0.5-4?g/ml
Type: Primary
Antigen: DCN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Pig, Rabbit, Rat