Order Entry
Slovakia
ContactUsLinkComponent
Fluorescein disodium salt 50 g/l in aqueous solution, VOLUSOL®
Fluorescein disodium salt 50 g/l in aqueous solution, VOLUSOL®
Catalog # VWRS83930.180
Supplier:  VWR Collection
CAS Number:  
undefined
Fluorescein disodium salt 50 g/l in aqueous solution, VOLUSOL®
Catalog # VWRS83930.180
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :376.28 g/mol
CAS Number:518-47-8
Formula:C₂₀H₁₀Na₂O₅
MDL Number:MFCD00167039