Filtering flask
Supplier: GLASSCO
Jump to:
About this item
Flasks are made of heat resistant borosilicate glass 3.3, therefore they are suitable for hot and cold filtration.
- Highly resistant against chemicals and thermal shock
- High temperature resistance
- With white inscription field
Due to the heavy wall thickness the VWR® filtration flask is vacuum tight.
2 Options Available Below
Product Details & Documents
Flasks are made of heat resistant borosilicate glass 3.3, therefore they are suitable for hot and cold filtration.
- Highly resistant against chemicals and thermal shock
- High temperature resistance
- With white inscription field
Due to the heavy wall thickness the VWR® filtration flask is vacuum tight.
Product Family Options
Product Information
- CapacityAvailabilityPrice
- GLAS259.245.04Vacuum filtering flask, for 25 mm membranes