Anti-TGFBR1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # USBIT8250-31A-APC
Supplier: UNITED STATES BIOLOGICAL
Specifications
- Antibody type:Primary
- Antigen name:transforming growth factor, beta receptor 1
- Antigen symbol:TGFBR1
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:internal
- Form:liquid
- Amino acid number:141 to 170
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: TGFBR1 (transforming growth factor, beta receptor 1)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse