Anti-MYC Mouse Monoclonal Antibody [clone: 9E10]
Catalog # USBIM9600-96Q
Supplier: UNITED STATES BIOLOGICAL
Specifications
- Antibody type:Primary
- Antigen name:v-myc avian myelocytomatosis viral oncogene homolog
- Antigen symbol:MYC
- Clonality:Monoclonal
- Clone:9E10
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Storage buffer:PBS, 15mM sodium azide, pH 7,4.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1-5 ?g/ml; Western Blot: 0.5-2?g/ml; Immunoprecipitation: 1-5 ?g/ml
Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: monoclonal
Clone: 90000000000
Conjugation: unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human