Anti-Mast Cell Protease 6 Goat Polyclonal Antibody
USBIM2414-50
- Antibody type:Primary
- Antigen name:Mast Cell Protease 6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Neutralisation:Yes
- Form:lyophilized powder
- Amino acid number:22 to 276
- Storage buffer:PBS, pH 7,4, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,2 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 0.1 ?g/ml; Immunoprecipitation: 25 ?g/ml
Type: Primary
Antigen: Mast Cell Protease 6
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse