Order Entry
Slovakia
ContactUsLinkComponent
Anti-HLA Class 1 Antigen G Mouse Monoclonal Antibody (Biotin) [clone: MEM-G/9]
Anti-HLA Class 1 Antigen G Mouse Monoclonal Antibody (Biotin) [clone: MEM-G/9]
Catalog # USBIH6098-72R1
Anti-HLA Class 1 Antigen G Mouse Monoclonal Antibody (Biotin) [clone: MEM-G/9]
Catalog # USBIH6098-72R1

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HLA Class 1 Antigen G
  • Clonality:
    Monoclonal
  • Clone:
    MEM-G/9
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 15mM sodium azide
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Flow Cytometry: 1:1000

Type: Primary
Antigen: HLA Class 1 Antigen G
Clonality: monoclonal
Clone: MEM-G/9
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human