Order Entry
Slovakia
ContactUsLinkComponent
Anti-GABA Receptor A, alpha 6 Rabbit polyclonal antibody
Anti-GABA Receptor A, alpha 6 Rabbit polyclonal antibody
Catalog # USBIG1015-21
Anti-GABA Receptor A, alpha 6 Rabbit polyclonal antibody
Catalog # USBIG1015-21

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    GABA A Receptor Alpha 6
  • Antigen symbol:
    GABRA6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Bovine,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    10mM HEPES, pH 7,5, 150mM sodium chloride,0,1mg/ml BSA, 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: GABA Receptor A, alpha 6
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Rat