Anti-Fibrin Degradation Product, D-Dimer Mouse Monoclonal Antibody [clone: 9L168]
USBIF4197-17C
- Antibody type:Primary
- Antigen name:Fibrin Degradation Product, D-Dimer
- Clonality:Monoclonal
- Clone:9L168
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~6mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: Fibrin Degradation Product, D-Dimer
Clonality: monoclonal
Clone: 9L168
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human