Anti-CCR4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 11D81]
Catalog # USBIC8350-01H
Supplier: UNITED BIOCHEMICALS
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) receptor 4
- Antigen symbol:CCR4
- Clonality:Monoclonal
- Clone:11D81
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Form:liquid
- Storage buffer:PBSup to 0,5% BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Do not freeze!
Type: Primary
Antigen: CCR4 (chemokine (C-C motif) receptor 4)
Clonality: Monoclonal
Clone: 11D81
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human