Order Entry
Slovakia
ContactUsLinkComponent
Anti-CKMT2 Rabbit Polyclonal Antibody (Biotin)
Anti-CKMT2 Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIC7910-21-BIOT
undefined
Anti-CKMT2 Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIC7910-21-BIOT

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Creatine Kinase, mitochondrial 2
  • Antigen symbol:
    CKMT2
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Type: Primary
Antigen: CKMT2 (creatine kinase, mitochondrial 2 (sarcomeric))
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse