Anti-Cdx-1/Cdx-2, Homeobox Protein Rabbit polyclonal antibody
USBIC2589-51-100
- Antibody type:Primary
- Antigen name:Homeobox Protein
- Antigen symbol:CDX1/2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rodent,Human
- Western blot:Yes
- Form:liquid
- Amino acid number:202 to 218
- Storage buffer:PBS, pH 7,2, 40% glycerol.
- Molecular weight:28.1 kDa
- Sequence:FQNRRAKERKVNKKKQQ
- Storage temperature:Cool
- Concentration:~0,05 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 1:2500
Type: Primary
Antigen: Cdx-1/Cdx-2, Homeobox Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rodent