Anti-CD53 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 3H1544]
USBIC2408-11D
- Antibody type:Primary
- Antigen name:CD53 molecule
- Antigen symbol:CD53
- Clonality:Monoclonal
- Clone:3H1544
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~100 Tests/1ml
- Shipping temperature:Ice
- Purification:purified by ion exchange chromatography
- Size:100 Tests
- Pk:100 Tests
Type: Primary
Antigen: CD53 (CD53 molecule)
Clonality: Monoclonal
Clone: 3H1544
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human