Anti-CD38 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: HIT2]
USBIC2390-20C
- Antibody type:Primary
- Antigen name:CD38 molecule
- Antigen symbol:CD38
- Clonality:Monoclonal
- Clone:HIT2
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage temperature:Cool
- Concentration:~4-5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:500 µl
- Pk:500 µl
Type: Primary
Antigen: CD38 (CD38 molecule)
Clonality: Monoclonal
Clone: HIT2
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human