Anti-CACNA1C Goat polyclonal antibody
Catalog # USBIC2097-85J
Supplier: UNITED STATES BIOLOGICAL
Specifications
- Antibody type:Primary
- Antigen name:calcium channel, voltage-dependent, L type, alpha 1C subunit
- Antigen symbol:CACNA1C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
- Sequence:C-RRRPSEEELQD
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:16000; Western Blot: 0.5-1. 5 ?g/ml
Type: Primary
Antigen: CACNA1C (calcium channel, voltage-dependent, L type, alpha 1C subunit)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human