Anti-Cardiolipin Human polyclonal antibody
USBIC1375
- Antibody type:Primary
- Antigen name:Cardiolipin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Human
- Isotype:IgG
- Reactivity:Human
- Form:lyophilized powder
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:2 ml
- Pk:2 mL
Recommended dilution: ELISA: 1:100
Type: Primary
Antigen: Cardiolipin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG
Reactivity: Human