Anti-Cadherin P Rabbit monoclonal antibody unconjugated [clone: 8A8]
USBIC0108-37B
- Antibody type:Primary
- Antigen name:Cadherin P
- Antigen symbol:CDHP
- Clonality:Monoclonal
- Clone:8A8
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Monkey,Human
- Western blot:Yes
- Storage buffer:10mMsodium HEPES, pH 7,5, 150 mM sodium chloride, 100ug/ml BSA, 50% glycerol, 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:1000; Immunofluorescence: 1:25
Type: Primary
Antigen: Cadherin P
Clonality: Monoclonal
Clone: 8A8
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey