Anti-ATF6 Rabbit Polyclonal Antibody
Catalog # USBI146077
Supplier: UNITED BIOCHEMICALS
Specifications
- Antibody type:Primary
- Antigen name:Activating transcription factor 6
- Antigen symbol:ATF6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
- Recommended dilution: Western Blot: 0.5-1 μg/ml; Immunochemistry: 10 μg/ml
Type: Primary
Antigen: ATF6 (activating transcription factor 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse