Anti-DUSP6 / MKP3 Goat Polyclonal Antibody
Catalog # EVBIEB11892
Supplier: EVEREST BIOTECH
Specifications
- Antibody type:Primary
- Antigen name:Dual specificity phosphatase 6
- Antigen symbol:DUSP6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Antigen synonyms:MKP3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: DUSP6 / MKP3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: