Anti-CLU Goat Polyclonal Antibody
Catalog # EVBIEB11646
Supplier: EVEREST BIOTECH
Specifications
- Antibody type:Primary
- Antigen name:clusterin
- Antigen symbol:CLU
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Amino acid number:44 to 58
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: CLU (clusterin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human