Anti-TRPC6 Goat Polyclonal Antibody
EVBIEB11508
- Antibody type:Primary
- Antigen name:transient receptor potential cation channel, subfamily C, member 6
- Antigen symbol:TRPC6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Rat,Mouse
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: TRPC6 (transient receptor potential cation channel, subfamily C, member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat