Anti-DBH Goat polyclonal antibody unconjugated
EVBIEB11321
- Antibody type:Primary
- Antigen name:Dopamine Beta Hydroxylase
- Antigen symbol:DBH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Amino acid number:437 to 448
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: DBH (dopamine beta-hydroxylase (dopamine beta-monooxygenase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: