Anti-MTHFR Goat Polyclonal Antibody
EVBIEB11165
- Antibody type:Primary
- Antigen name:Methylenetetrahydrofolate reductase (NADPH)
- Antigen symbol:MTHFR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Amino acid number:369 to 383
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: MTHFR (methylenetetrahydrofolate reductase (NAD(P)H))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: