Anti-VPREB3 Goat Polyclonal Antibody
Catalog # EVBIEB09713
Supplier: EVEREST BIOTECH
Specifications
- Antibody type:Primary
- Antigen name:pre-B lymphocyte 3
- Antigen symbol:VPREB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Pig,Human,Rat,Mouse
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: VPREB3 (pre-B lymphocyte 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Pig, Rat