Order Entry
Slovakia
ContactUsLinkComponent
Anti-KIAA0319 Goat Polyclonal Antibody
Anti-KIAA0319 Goat Polyclonal Antibody
Catalog # EVBIEB07060
Supplier:  EVEREST BIOTECH
Anti-KIAA0319 Goat Polyclonal Antibody
Catalog # EVBIEB07060
Supplier:  EVEREST BIOTECH

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    KIAA0319
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Epitope:
    internal
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: KIAA0319 (KIAA0319)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human