Anti-AKR1C3 Goat Polyclonal Antibody
Catalog # EVBIEB06637
Supplier: EVEREST BIOTECH
Specifications
- Antibody type:Primary
- Antigen name:aldo-keto reductase family 1, member C3
- Antigen symbol:AKR1C3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: AKR1C3 (aldo-keto reductase family 1, member C3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human