Anti-CCDC50 Goat Polyclonal Antibody
EVBIEB06551
- Antibody type:Primary
- Antigen name:coiled-coil domain containing 50
- Antigen symbol:CCDC50
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: CCDC50 (coiled-coil domain containing 50)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: