Anti-LRRK2 Goat Polyclonal Antibody
Catalog # EVBIEB06550
Supplier: EVEREST BIOTECH
Specifications
- Antibody type:Primary
- Antigen name:leucine-rich repeat kinase 2
- Antigen symbol:LRRK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Bovine
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: LRRK2 (leucine-rich repeat kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Bovine, Human